Product Details

SNP ID
rs1262131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59453321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTGGTAACAGGTGAGGTTTTGC[A/T]TGTGTTGGGGCAAAGAATAAATGGG
Phenotype
MIM: 607970
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR135 PubMed Links

Gene Details

Gene
GPR135
Gene Name
G protein-coupled receptor 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022571.5 7666 Intron NP_072093.2
XM_017021596.1 7666 UTR 3 XP_016877085.1
XM_017021597.1 7666 UTR 3 XP_016877086.1
XM_017021598.1 7666 UTR 3 XP_016877087.1
XM_017021599.1 7666 UTR 3 XP_016877088.1

View Full Product Details