Product Details

SNP ID
rs13238458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87881104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATGTGCACTGCACTCTGTGTTTT[A/G]GTAGATTCAAAAATGAATCCAGCCA
Phenotype
MIM: 604281 MIM: 610821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBF4 PubMed Links

Gene Details

Gene
DBF4
Gene Name
DBF4 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318060.1 Intron NP_001304989.1
NM_001318061.1 Intron NP_001304990.1
NM_001318062.1 Intron NP_001304991.1
NM_006716.3 Intron NP_006707.1
Gene
SLC25A40
Gene Name
solute carrier family 25 member 40
There are no transcripts associated with this gene.

View Full Product Details