Product Details

SNP ID
rs1472146
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:128863888 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACCTGGGAGGCTGTTATCTGCCA[A/T]TCTCTAAGCTGTCTGGTAAAGTGAC
Phenotype
MIM: 607160 MIM: 102565 MIM: 609344
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1F PubMed Links

Gene Details

Gene
ATP6V1F
Gene Name
ATPase H+ transporting V1 subunit F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198909.1 Intron NP_001185838.1
NM_004231.3 Intron NP_004222.2
Gene
FLNC
Gene Name
filamin C
There are no transcripts associated with this gene.

Gene
KCP
Gene Name
kielin/chordin-like protein
There are no transcripts associated with this gene.

Gene
LOC100130705
Gene Name
uncharacterized LOC100130705
There are no transcripts associated with this gene.

View Full Product Details