Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001005609.1 | 880 | Intron | NP_001005609.1 | ||
NM_001005610.3 | 880 | Intron | NP_001005610.2 | ||
NM_001005612.2 | 880 | Intron | NP_001005612.2 | ||
NM_001005613.3 | 880 | Intron | NP_001005613.1 | ||
NM_001399.4 | 880 | Intron | NP_001390.1 | ||
XM_006724630.2 | 880 | Intron | XP_006724693.1 | ||
XM_011530885.2 | 880 | Intron | XP_011529187.1 | ||
XM_017029336.1 | 880 | Intron | XP_016884825.1 | ||
XM_017029337.1 | 880 | UTR 3 | XP_016884826.1 |