Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007792.1 | Intron | NP_001007793.1 | |||
NM_001012331.1 | Intron | NP_001012331.1 | |||
NM_002529.3 | Intron | NP_002520.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161441.1 | Intron | NP_001154913.1 | |||
NM_001161442.1 | Intron | NP_001154914.1 | |||
NM_001161443.1 | Intron | NP_001154915.1 | |||
NM_001161444.1 | Intron | NP_001154916.1 | |||
NM_003975.3 | Intron | NP_003966.2 | |||
XM_006711615.2 | Intron | XP_006711678.1 | |||
XM_017002762.1 | Intron | XP_016858251.1 | |||
XM_017002763.1 | Intron | XP_016858252.1 | |||
XM_017002764.1 | Intron | XP_016858253.1 | |||
XM_017002765.1 | Intron | XP_016858254.1 | |||
XM_017002766.1 | Intron | XP_016858255.1 | |||
XM_017002767.1 | Intron | XP_016858256.1 |