Product Details

SNP ID
rs7042087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129840589 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACCTAGATTCACGAATTAGTAA[C/T]ATTTGCCTTATTTACTCTAACTCTA
Phenotype
MIM: 615143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf78 PubMed Links

Gene Details

Gene
C9orf78
Gene Name
chromosome 9 open reading frame 78
There are no transcripts associated with this gene.

Gene
USP20
Gene Name
ubiquitin specific peptidase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008563.4 Intron NP_001008563.2
NM_001110303.3 Intron NP_001103773.2
NM_006676.7 Intron NP_006667.3
XM_005251665.3 Intron XP_005251722.1
XM_005251666.3 Intron XP_005251723.1
XM_011518161.2 Intron XP_011516463.1
XM_011518162.2 Intron XP_011516464.1
XM_017014221.1 Intron XP_016869710.1

View Full Product Details