Product Details

SNP ID
rs28458971
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:133389952 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCCTGCTGAGCCTTTGACCCCCAG[C/T]GGCACAACTTTCAGGCTGGAGAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STKLD1 PubMed Links

Gene Details

Gene
STKLD1
Gene Name
serine/threonine kinase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153710.4 Intron NP_714921.4

View Full Product Details