Product Details

SNP ID
rs16933086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36215584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCTCTCTAGCCCCATAGCTTAA[C/T]AGTCAAACACACCAAGTTCTATAAA
Phenotype
MIM: 118960 MIM: 603824
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLTA PubMed Links

Gene Details

Gene
CLTA
Gene Name
clathrin light chain A
There are no transcripts associated with this gene.

Gene
GNE
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128227.2 4016 UTR 3 NP_001121699.1
NM_001190383.1 4016 UTR 3 NP_001177312.1
NM_001190384.1 4016 UTR 3 NP_001177313.1
NM_001190388.1 4016 UTR 3 NP_001177317.1
NM_005476.5 4016 UTR 3 NP_005467.1
XM_005251334.4 4016 Intron XP_005251391.1
XM_017014167.1 4016 UTR 3 XP_016869656.1
XM_017014168.1 4016 UTR 3 XP_016869657.1

View Full Product Details