Product Details

SNP ID
rs4898989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59468626 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAACCATGGTCTATAATTTTTTGC[A/G]TGATTATCTAAGCATTACCTGCCTC
Phenotype
MIM: 607970 MIM: 614811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR135 PubMed Links

Gene Details

Gene
GPR135
Gene Name
G protein-coupled receptor 135
There are no transcripts associated with this gene.

Gene
L3HYPDH
Gene Name
trans-L-3-hydroxyproline dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144581.1 Intron NP_653182.1
XM_005267298.3 Intron XP_005267355.1
XM_011536386.2 Intron XP_011534688.1
XM_011536387.1 Intron XP_011534689.1
XM_011536388.1 Intron XP_011534690.1

View Full Product Details