Product Details

SNP ID
rs11078780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2030905 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATAGCAGTGGTAACGTTCAAATC[G/T]GATCTCAGGGCCCCAGCCTCCGCTG
Phenotype
MIM: 603527 MIM: 610461
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPH1 PubMed Links

Gene Details

Gene
DPH1
Gene Name
diphthamide biosynthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001383.3 Intron NP_001374.3
Gene
RTN4RL1
Gene Name
reticulon 4 receptor like 1
There are no transcripts associated with this gene.

View Full Product Details