Product Details
- SNP ID
-
rs746677
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:86940881 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CACAGAGCCTAGTTGTCCCCCTCCC[A/G]GGACCACACTCAGGGAGGGGATGGG
- Phenotype
-
MIM: 608925
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MMRN2
PubMed Links
Gene Details
- Gene
- MMRN2
- Gene Name
- multimerin 2
View Full Product Details