Product Details

SNP ID
rs11862450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85903427 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTATTCTGATGAGGAGATCGAAG[C/G]CTCTTTTCTTCTCTTTTCTGCTGCT
Phenotype
MIM: 601565
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRF8 PubMed Links

Gene Details

Gene
IRF8
Gene Name
interferon regulatory factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002163.2 Intron NP_002154.1
XM_017023199.1 Intron XP_016878688.1

View Full Product Details