Product Details
- SNP ID
-
rs12830005
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48981893 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTCCCTCCCTCTCCGCGGGAGA[A/C]CCCTTGTTGCACTGCCCCCTGCTTG
- Phenotype
-
MIM: 164820
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WNT1
PubMed Links
Gene Details
- Gene
- WNT1
- Gene Name
- Wnt family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005430.3 |
1564 |
UTR 3 |
|
|
NP_005421.1 |
View Full Product Details