Product Details

SNP ID
rs11952545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36103149 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACCTTTGAATAACCAGTTTTTT[C/T]CTCAAAAAGTATAAGCAGGTACCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMBRD2 PubMed Links

Gene Details

Gene
LMBRD2
Gene Name
LMBR1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007527.1 3842 Intron NP_001007528.1
XM_011514162.2 3842 UTR 3 XP_011512464.1
XM_017010024.1 3842 UTR 3 XP_016865513.1

View Full Product Details