Product Details
- SNP ID
-
rs7554597
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:177932523 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACAAGCCTCCAACCCCAGCGCC[C/T]GCAGCTGCTCCCCCGCTGGATGCGG
- Phenotype
-
MIM: 612855
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SEC16B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3748792] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEC16B
- Gene Name
- SEC16 homolog B, endoplasmic reticulum export factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033127.2 |
3069 |
Silent Mutation |
GCA,GCG |
A993A |
NP_149118.2 |
View Full Product Details