Product Details

SNP ID
rs2643195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39696865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGGGGAGAAAGGACACCTCT[A/G]ACCCTGATCCTAACATCCCGATGGC
Phenotype
MIM: 164870 MIM: 611801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERBB2 PubMed Links

Gene Details

Gene
ERBB2
Gene Name
erb-b2 receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005862.2 Intron NP_001005862.1
NM_001289936.1 Intron NP_001276865.1
NM_001289937.1 Intron NP_001276866.1
NM_001289938.1 Intron NP_001276867.1
NM_004448.3 Intron NP_004439.2
Gene
PGAP3
Gene Name
post-GPI attachment to proteins 3
There are no transcripts associated with this gene.

View Full Product Details