Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001100412.1 | 4985 | UTR 3 | NP_001093882.1 | ||
NM_013235.4 | 4985 | UTR 3 | NP_037367.3 | ||
XM_005248291.3 | 4985 | Intron | XP_005248348.1 | ||
XM_005248292.3 | 4985 | Intron | XP_005248349.1 | ||
XM_005248293.3 | 4985 | Intron | XP_005248350.1 | ||
XM_005248294.3 | 4985 | Intron | XP_005248351.1 | ||
XM_011514033.2 | 4985 | Intron | XP_011512335.1 | ||
XM_017009399.1 | 4985 | Intron | XP_016864888.1 | ||
XM_017009400.1 | 4985 | Intron | XP_016864889.1 | ||
XM_017009401.1 | 4985 | Intron | XP_016864890.1 | ||
XM_017009402.1 | 4985 | Intron | XP_016864891.1 |