Product Details
- SNP ID
-
rs8106096
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18425606 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTAGACAGGTGGGCCTGCAGGCCGC[C/G]GGGCCAGGAAAGTGGAGGAAGAGCA
- Phenotype
-
MIM: 611670
MIM: 607391
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ISYNA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150781536] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ISYNA1
- Gene Name
- inositol-3-phosphate synthase 1
There are no transcripts associated with this gene.
- Gene
- SSBP4
- Gene Name
- single stranded DNA binding protein 4
View Full Product Details