Product Details

SNP ID
rs12563094
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:109409923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAATCCAACAATAAAACAGAA[A/G]GTACCTCAATGTTTGTTGCATTCAG
Phenotype
MIM: 176844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMA5 PubMed Links

Gene Details

Gene
PSMA5
Gene Name
proteasome subunit alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199772.1 Intron NP_001186701.1
NM_001199773.1 Intron NP_001186702.1
NM_001199774.1 Intron NP_001186703.1
NM_002790.3 Intron NP_002781.2

View Full Product Details