Product Details
- SNP ID
-
rs475820
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:42745306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGAAAAAAAACCTTCATTATTTT[A/C]AGCTGTTGACATTTTGCAATTGTTT
- Phenotype
-
MIM: 609592
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RIT2
PubMed Links
Gene Details
- Gene
- RIT2
- Gene Name
- Ras like without CAAX 2
View Full Product Details