Product Details
- SNP ID
-
rs10739289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97032472 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTGGTAGATTCAGGAGTTGGGT[A/T]ACACACAGTAACTGGGCAGCTGATA
- Phenotype
-
MIM: 603308
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTSV
PubMed Links
Gene Details
- Gene
- CTSV
- Gene Name
- cathepsin V
View Full Product Details