Product Details

SNP ID
rs3824930
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102114571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGTTTGGTATGACCTAAATAC[C/T]TTTGTTAGTATGGGTAATATTTTGA
Phenotype
MIM: 606608
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
YAP1 PubMed Links

Gene Details

Gene
YAP1
Gene Name
Yes associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130145.2 Intron NP_001123617.1
NM_001195044.1 Intron NP_001181973.1
NM_001195045.1 Intron NP_001181974.1
NM_001282097.1 Intron NP_001269026.1
NM_001282098.1 Intron NP_001269027.1
NM_001282099.1 Intron NP_001269028.1
NM_001282100.1 Intron NP_001269029.1
NM_001282101.1 Intron NP_001269030.1
NM_006106.4 Intron NP_006097.2
XM_005271378.3 Intron XP_005271435.1
XM_005271380.3 Intron XP_005271437.1
XM_005271381.3 Intron XP_005271438.1
XM_005271383.3 Intron XP_005271440.1
XM_011542555.2 Intron XP_011540857.1
XM_011542556.2 Intron XP_011540858.1
XM_017017093.1 Intron XP_016872582.1

View Full Product Details