Product Details

SNP ID
rs2255405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6606823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCCCCGAGTTTGGCCTGCCCT[A/G]GCGCCACAGGCAGTACTGGAAGCTG
Phenotype
MIM: 602366 MIM: 615818 MIM: 600475 MIM: 607998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILK PubMed Links

Gene Details

Gene
ILK
Gene Name
integrin linked kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014794.2 Intron NP_001014794.1
NM_001014795.2 Intron NP_001014795.1
NM_001278441.1 Intron NP_001265370.1
NM_001278442.1 Intron NP_001265371.1
NM_004517.3 Intron NP_004508.1
XM_005252904.4 Intron XP_005252961.1
XM_005252905.2 Intron XP_005252962.1
XM_011520065.1 Intron XP_011518367.1
XM_017017672.1 Intron XP_016873161.1
Gene
RRP8
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
There are no transcripts associated with this gene.

Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
There are no transcripts associated with this gene.

Gene
TPP1
Gene Name
tripeptidyl peptidase 1
There are no transcripts associated with this gene.

View Full Product Details