Product Details

SNP ID
rs287987
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:9846033 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGTTGGGAATATTGGAATTATTCTT[C/T]GTTATCCTTTTACCTTTGTATAGTA
Phenotype
MIM: 604904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF1B PubMed Links

Gene Details

Gene
TAF1B
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318976.1 Intron NP_001305905.1
NM_001318977.1 Intron NP_001305906.1
NM_005680.2 Intron NP_005671.2
XM_011510410.2 Intron XP_011508712.1
XM_011510411.1 Intron XP_011508713.1

View Full Product Details