Product Details

SNP ID
rs385513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9843587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGCGGTGCACCTGGCGGGCC[A/C]CGATCGCCGGGGCCGGAGGAGAGAG
Phenotype
MIM: 604904
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAF1B PubMed Links

Gene Details

Gene
TAF1B
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318976.1 Intron NP_001305905.1
NM_001318977.1 Intron NP_001305906.1
NM_005680.2 Intron NP_005671.2
XM_011510410.2 Intron XP_011508712.1
XM_011510411.1 Intron XP_011508713.1

View Full Product Details