Product Details

SNP ID
rs13013695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32166162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTTCAGCTCCTGGCGAGTTTCT[A/G]ACTTCCCTCTGCCGAGATTTCTTTA
Phenotype
MIM: 611148 MIM: 604277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A6 PubMed Links

Gene Details

Gene
SLC30A6
Gene Name
solute carrier family 30 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006712048.3 292 Intron XP_006712111.1
XM_011532959.2 292 Intron XP_011531261.1
XM_011532960.2 292 UTR 5 XP_011531262.1
XM_011532961.2 292 Intron XP_011531263.1
XM_011532962.2 292 UTR 5 XP_011531264.1
XM_011532963.2 292 Intron XP_011531265.1
XM_017004462.1 292 Intron XP_016859951.1
XM_017004463.1 292 Intron XP_016859952.1
XM_017004464.1 292 Intron XP_016859953.1
XM_017004465.1 292 Intron XP_016859954.1
XM_017004466.1 292 Intron XP_016859955.1
Gene
SPAST
Gene Name
spastin
There are no transcripts associated with this gene.

View Full Product Details