Product Details

SNP ID
rs2280968
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:32064971 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTAAAACCTAAAACAACTATGTAT[G/T]TTTCTGAATGAAAGCAAGGATACTA
Phenotype
MIM: 604277
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPAST PubMed Links

Gene Details

Gene
SPAST
Gene Name
spastin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014946.3 Intron NP_055761.2
NM_199436.1 Intron NP_955468.1
XM_005264516.4 Intron XP_005264573.1
XM_011533067.2 Intron XP_011531369.1
XM_017004777.1 Intron XP_016860266.1
XM_017004778.1 Intron XP_016860267.1

View Full Product Details