Product Details
- SNP ID
-
rs6758024
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32069417 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAATGTAATTCTTCCTCTTTCTCA[C/G]TGCTTCAAAATATATATGATCAAAT
- Phenotype
-
MIM: 604277
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPAST
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62142107] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPAST
- Gene Name
- spastin
View Full Product Details