Product Details

SNP ID
rs615896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75564256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCAGGAGCAGGCGCTAAGCTGC[C/T]GCGGGCCTAGCGGGCTGCCTGAGGA
Phenotype
MIM: 600943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINH1 PubMed Links

Gene Details

Gene
SERPINH1
Gene Name
serpin family H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207014.1 Intron NP_001193943.1
NM_001235.3 Intron NP_001226.2
XM_011545327.1 Intron XP_011543629.1

View Full Product Details