Product Details
- SNP ID
-
rs778584
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:140625627 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGCACTTACCATCATCTAGCACT[C/T]TATACTTTCTTCTCCCCACTGCAAT
- Phenotype
-
MIM: 158120
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78947429] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD14
- Gene Name
- CD14 molecule
There are no transcripts associated with this gene.
- Gene
- TMCO6
- Gene Name
- transmembrane and coiled-coil domains 6
View Full Product Details