Product Details
- SNP ID
-
rs296890
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:83980816 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGGTAAAAAAGGAACTGGGAGCG[A/C]GCCGCGGCGGTTCCTGTCCTTACAG
- Phenotype
-
MIM: 600712
MIM: 610404
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs188055179] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNRNPK
- Gene Name
- heterogeneous nuclear ribonucleoprotein K
- Gene
- RMI1
- Gene Name
- RecQ mediated genome instability 1
View Full Product Details