Product Details

SNP ID
rs2917549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103376190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAACCATAGTATCTAAAATAAC[A/G]TACTTTTTAAAAATGCAAACAAAAG
Phenotype
MIM: 610635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTHRC1 PubMed Links

Gene Details

Gene
CTHRC1
Gene Name
collagen triple helix repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256099.1 Intron NP_001243028.1
NM_138455.3 Intron NP_612464.1
XM_011516824.2 Intron XP_011515126.1

View Full Product Details