Product Details
- SNP ID
-
rs13034933
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32016700 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTATTCCTGATTCAATCTACTGAT[A/C]CTCCCAACTAACCCACCCACCAACG
- Phenotype
-
MIM: 612032
MIM: 611786
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DPY30
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150065663] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPY30
- Gene Name
- dpy-30, histone methyltransferase complex regulatory subunit
- Gene
- MEMO1
- Gene Name
- mediator of cell motility 1
There are no transcripts associated with this gene.
View Full Product Details