Product Details
- SNP ID
-
rs13079063
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52710444 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGTGTGTGTGTAATTTTCAAAG[A/G]TAGATATATTTTTAGTTATAGAAAC
- Phenotype
-
MIM: 601959
MIM: 610358
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLT8D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149305355] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLT8D1
- Gene Name
- glycosyltransferase 8 domain containing 1
There are no transcripts associated with this gene.
- Gene
- NEK4
- Gene Name
- NIMA related kinase 4
- Gene
- SPCS1
- Gene Name
- signal peptidase complex subunit 1
There are no transcripts associated with this gene.
View Full Product Details