Product Details

SNP ID
rs12424158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:67273205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTGGGACAGTACCTTAGAACAG[C/T]GCTTCTCATCACTTCCTCCCTAAAA
Phenotype
MIM: 607727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAND1 PubMed Links

Gene Details

Gene
CAND1
Gene Name
cullin associated and neddylation dissociated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018448.4 Intron NP_060918.2

View Full Product Details