Product Details

SNP ID
rs34603458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199933729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATTACTTGAGGAGTATCTTTG[A/T]CCTTTTCTGATTCATTATAATACTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf69 PubMed Links

Gene Details

Gene
C2orf69
Gene Name
chromosome 2 open reading frame 69
There are no transcripts associated with this gene.

Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039693.2 Intron NP_001034782.1
XM_017003310.1 Intron XP_016858799.1

View Full Product Details