Product Details

SNP ID
rs12917341
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:92624570 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCTGCAGAGGCCTCATGCTGCAAAC[G/T]GCACAAAAATGCCCTTTGTTTCCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM174B PubMed Links

Gene Details

Gene
FAM174B
Gene Name
family with sequence similarity 174 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207446.2 Intron NP_997329.2
Gene
LOC105370978
Gene Name
uncharacterized LOC105370978
There are no transcripts associated with this gene.

View Full Product Details