Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_025260.3 | 185 | Intron | NP_079536.2 | ||
NM_138272.2 | 185 | Intron | NP_612116.1 | ||
NM_138273.2 | 185 | Intron | NP_612117.1 | ||
NM_138274.2 | 185 | Intron | NP_612118.1 | ||
NM_138275.2 | 185 | Intron | NP_612119.1 | ||
NM_138277.2 | 185 | Intron | NP_612121.1 | ||
XM_011514920.2 | 185 | Intron | XP_011513222.1 | ||
XM_011514921.2 | 185 | Intron | XP_011513223.1 | ||
XM_017011330.1 | 185 | Intron | XP_016866819.1 | ||
XM_017011331.1 | 185 | Intron | XP_016866820.1 | ||
XM_017011332.1 | 185 | Intron | XP_016866821.1 | ||
XM_017011333.1 | 185 | UTR 5 | XP_016866822.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_025261.2 | 185 | Intron | NP_079537.1 |