Product Details

SNP ID
rs1061485
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:25926633 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACTCATTCCTTTATTGTCAACTGC[A/G]TTGATTGGAACAGGGCTGGGGCCTG
Phenotype
MIM: 602845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF3C PubMed Links

Gene Details

Gene
KIF3C
Gene Name
kinesin family member 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002254.6 5381 UTR 3 NP_002245.4
XM_005264299.2 5381 UTR 3 XP_005264356.2

View Full Product Details