Product Details

SNP ID
rs11536845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135481186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAATTTGACAAAGCCTGCACCT[C/G]TGGCTTGGGCCCACACCTATCTCTA
Phenotype
MIM: 614056
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R26 PubMed Links

Gene Details

Gene
PPP1R26
Gene Name
protein phosphatase 1 regulatory subunit 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014811.3 Intron NP_055626.3
XM_005263411.2 Intron XP_005263468.1
XM_017015355.1 Intron XP_016870844.1
XM_017015356.1 Intron XP_016870845.1
XM_017015357.1 Intron XP_016870846.1
XM_017015358.1 Intron XP_016870847.1
XM_017015359.1 Intron XP_016870848.1
XM_017015360.1 Intron XP_016870849.1
XM_017015361.1 Intron XP_016870850.1
XM_017015362.1 Intron XP_016870851.1
Gene
PPP1R26-AS1
Gene Name
PPP1R26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details