Product Details

SNP ID
rs11722455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2795756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGAGGGTGGGAGACCAGTTTGA[C/T]GGGAGTGGCCTTTGCATGCCAGGTG
Phenotype
MIM: 602104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3BP2 PubMed Links

Gene Details

Gene
SH3BP2
Gene Name
SH3 domain binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122681.1 Intron NP_001116153.1
NM_001145855.1 Intron NP_001139327.1
NM_001145856.1 Intron NP_001139328.1
NM_003023.4 Intron NP_003014.3

View Full Product Details