Product Details

SNP ID
rs9368923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36355764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAACTAGAATCATATCTAAAT[A/G]TCAGGCCTTTAATCCCTGTAGGAGG
Phenotype
MIM: 605255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETV7 PubMed Links

Gene Details

Gene
ETV7
Gene Name
ETS variant 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207035.1 Intron NP_001193964.1
NM_001207036.1 Intron NP_001193965.1
NM_001207037.1 Intron NP_001193966.1
NM_001207038.1 Intron NP_001193967.1
NM_001207039.1 Intron NP_001193968.1
NM_001207040.1 Intron NP_001193969.1
NM_001207041.1 Intron NP_001193970.1
NM_016135.3 Intron NP_057219.1
XM_011514659.1 Intron XP_011512961.1

View Full Product Details