Product Details

SNP ID
rs6088635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34878698 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGAATGCATTTCACCAAATTCC[A/G]TCAAATACAGAATTTCTACCAGGGA
Phenotype
MIM: 605832 MIM: 612342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSS2 PubMed Links

Gene Details

Gene
ACSS2
Gene Name
acyl-CoA synthetase short-chain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076552.2 Intron NP_001070020.2
NM_001242393.1 Intron NP_001229322.1
NM_018677.3 Intron NP_061147.1
XM_005260455.1 Intron XP_005260512.1
XM_005260456.1 Intron XP_005260513.1
XM_006723826.1 Intron XP_006723889.1
XM_011528905.1 Intron XP_011527207.1
XM_011528906.1 Intron XP_011527208.1
XM_011528907.1 Intron XP_011527209.1
XM_011528908.1 Intron XP_011527210.1
XM_011528909.2 Intron XP_011527211.1
XM_011528911.1 Intron XP_011527213.1
XM_011528912.2 Intron XP_011527214.1
Gene
GGT7
Gene Name
gamma-glutamyltransferase 7
There are no transcripts associated with this gene.

View Full Product Details