Product Details

SNP ID
rs2256319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4724311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCTTTGCCATTCTTACCTGTCT[A/C]GGAAACCGGGACCAGAGCCCTGAGT
Phenotype
MIM: 604263
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRND PubMed Links

Gene Details

Gene
PRND
Gene Name
prion protein 2 (dublet)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012409.3 Intron NP_036541.2
Gene
PRNT
Gene Name
prion protein (testis specific)
There are no transcripts associated with this gene.

View Full Product Details