Product Details

SNP ID
rs6495498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80419287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATGCTGGTACTGCTGGCCCAGAT[C/G]CCCTTCAGGTCCACTGAGGCAGGGG
Phenotype
MIM: 606036
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARNT2 PubMed Links

Gene Details

Gene
ARNT2
Gene Name
aryl hydrocarbon receptor nuclear translocator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014862.3 Intron NP_055677.3

View Full Product Details