Product Details

SNP ID
rs10991598
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:105249075 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAAACTGCCGCTTGTCTGTCTAGC[G/T]TTAGGAGACATTTGTCCACTGTCTC
Phenotype
MIM: 606105
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC44A1 PubMed Links

Gene Details

Gene
SLC44A1
Gene Name
solute carrier family 44 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286730.1 Intron NP_001273659.1
NM_080546.4 Intron NP_536856.2
XM_005251855.4 Intron XP_005251912.1
XM_006717027.3 Intron XP_006717090.1
XM_006717028.3 Intron XP_006717091.1
XM_006717029.2 Intron XP_006717092.1
XM_017014559.1 Intron XP_016870048.1
XM_017014560.1 Intron XP_016870049.1

View Full Product Details