Product Details
- SNP ID
-
rs711822
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:176124295 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCAGCGACTTGGACTTCTTCTT[T/C]TTTTTTTTTTTCTTTTTAGATAGAA
- Phenotype
-
MIM: 142984
MIM: 142985
MIM: 142982
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HOXD10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62188694] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HOXD10
- Gene Name
- homeobox D10
There are no transcripts associated with this gene.
- Gene
- HOXD8
- Gene Name
- homeobox D8
There are no transcripts associated with this gene.
- Gene
- HOXD9
- Gene Name
- homeobox D9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014213.3 |
1263 |
UTR 3 |
|
|
NP_055028.3 |
- Gene
- LOC100129455
- Gene Name
- uncharacterized LOC100129455
There are no transcripts associated with this gene.
View Full Product Details