Product Details

SNP ID
rs7598552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108323614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGCCAGGGCATCTATTAGGCAGT[C/T]GAATGCATTGAAGGGGGTCAAAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1C2P1 PubMed Links

Gene Details

Gene
SULT1C2P1
Gene Name
sulfotransferase family 1C member 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details