Product Details

SNP ID
rs11689073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:117815820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTCAGGTACAGTCATGCATCGC[A/G]TAACAGGAGGGGGGATACATTGTGA
Phenotype
MIM: 606355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX18 PubMed Links

Gene Details

Gene
DDX18
Gene Name
DEAD-box helicase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006773.3 Intron NP_006764.3

View Full Product Details