Product Details

SNP ID
rs2516801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167996390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACAGCGCAGCCTGGGTCCAAAGC[A/G]GTGAGGAGCACAGGCCACAGCACGC
Phenotype
MIM: 603815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF25 PubMed Links

Gene Details

Gene
KIF25
Gene Name
kinesin family member 25
There are no transcripts associated with this gene.

Gene
KIF25-AS1
Gene Name
KIF25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details